Product Details
- SNP ID
-
rs188640709
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:97994693 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGGAAAAATTGAGGGCTTAACAGC[A/T]GAATTTGTGAACTTAGAGTTCCTCA
- Phenotype
-
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ANP32B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs562853890] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ANP32B
- Gene Name
- acidic nuclear phosphoprotein 32 family member B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006401.2 |
466 |
Silent Mutation |
GCA,GCT |
A39A |
NP_006392.1 |
View Full Product Details