Product Details

SNP ID
rs189410619
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42851505 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGTGCCCAGCCTTGGACACACCC[C/T]GGCCAGAGCCAGCTGTTTGCAGACC
Phenotype
MIM: 602268 MIM: 603735 MIM: 605129
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AOC2 PubMed Links

Gene Details

Gene
AOC2
Gene Name
amine oxidase, copper containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001158.3 383 Intron NP_001149.2
NM_009590.2 383 Intron NP_033720.2
Gene
AOC3
Gene Name
amine oxidase, copper containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277731.1 383 Silent Mutation CCC,CCT P54P NP_001264660.1
NM_001277732.1 383 Intron NP_001264661.1
NM_003734.3 383 Silent Mutation CCC,CCT P54P NP_003725.1
XM_011525419.1 383 Silent Mutation CCC,CCT P54P XP_011523721.1
XM_011525420.2 383 Silent Mutation CCC,CCT P54P XP_011523722.1
Gene
PSME3
Gene Name
proteasome activator subunit 3
There are no transcripts associated with this gene.

View Full Product Details