Product Details

SNP ID
rs189051075
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:112230230 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTTGTGGTGACAGTACATGGAGA[A/G]GTATGTGCAGAAAATATTTGTGTGG
Phenotype
MIM: 612719
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTS PubMed Links

Gene Details

Gene
PTS
Gene Name
6-pyruvoyltetrahydropterin synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000317.2 265 Silent Mutation GAA,GAG E62E NP_000308.1

View Full Product Details