Product Details

SNP ID
rs189201910
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81941103 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCTGTGTGGCCCATCACACTCAG[A/G]GCCACCACGGCCACTGTGGCCAGGA
Phenotype
MIM: 179035
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYADML2 PubMed Links

Gene Details

Gene
MYADML2
Gene Name
myeloid associated differentiation marker like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145113.2 994 Silent Mutation GCC,GCT A213A NP_001138585.2
Gene
PYCR1
Gene Name
pyrroline-5-carboxylate reductase 1
There are no transcripts associated with this gene.

View Full Product Details