Product Details

SNP ID
rs189234701
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:5867458 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCATCTCTGAGCAGCGGGGCAGAG[C/T]CACGGGTGTGTTAAAAGGCCACGTC
Phenotype
MIM: 136835
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FUT5 PubMed Links

Gene Details

Gene
FUT5
Gene Name
fucosyltransferase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002034.2 356 Missense Mutation NP_002025.2
Gene
LOC101928844
Gene Name
uncharacterized LOC101928844
There are no transcripts associated with this gene.

View Full Product Details