Product Details

SNP ID
rs189173553
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:80485851 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGTCCCAGGTGAACTCGTGGCGA[C/G]AGAAGGAAAATGCGCGTCCCGGGCG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZBTB10 PubMed Links

Gene Details

Gene
ZBTB10
Gene Name
zinc finger and BTB domain containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105539.2 233 Intron NP_001099009.1
NM_001277145.1 233 Missense Mutation ACA,AGA T23R NP_001264074.1
NM_023929.4 233 Intron NP_076418.3

View Full Product Details