Product Details

SNP ID
rs189510894
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:8842844 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATGAGGTCAGATACCATCAATGG[C/G]AGGAGAGCTTGGAGAAGCCTCAGCA
Phenotype
MIM: 607963 MIM: 606154
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MBD3L1 PubMed Links

Gene Details

Gene
MBD3L1
Gene Name
methyl-CpG binding domain protein 3 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145208.2 252 Missense Mutation CAG,GAG Q56E NP_660209.2
Gene
MUC16
Gene Name
mucin 16, cell surface associated
There are no transcripts associated with this gene.

View Full Product Details