Product Details

SNP ID
rs189549581
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:33387460 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTCTATGCCGCTGTCAGTCACCA[C/T]GACGTCCTCCTCGATGCGGACCTGG
Phenotype
MIM: 138972 MIM: 613230
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CEBPG PubMed Links

Gene Details

Gene
CEBPG
Gene Name
CCAAT/enhancer binding protein gamma
There are no transcripts associated with this gene.

Gene
PEPD
Gene Name
peptidase D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000285.3 1376 Missense Mutation ATG,GTG M456V NP_000276.2
NM_001166056.1 1376 Missense Mutation ATG,GTG M415V NP_001159528.1
NM_001166057.1 1376 Missense Mutation ATG,GTG M392V NP_001159529.1

View Full Product Details