Product Details

SNP ID
rs189981520
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30525530 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGGCAGGTCCCCTGTGGGCTGCT[C/T]CCCAAACCCCTGAGTGAAGGAGTCC
Phenotype
MIM: 153370
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ITGAL PubMed Links

Gene Details

Gene
ITGAL
Gene Name
integrin subunit alpha L
There are no transcripts associated with this gene.

Gene
ZNF747
Gene Name
zinc finger protein 747
There are no transcripts associated with this gene.

Gene
ZNF768
Gene Name
zinc finger protein 768
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024671.3 611 Missense Mutation AAG,GAG K204E NP_078947.3
XM_017023665.1 611 Missense Mutation AAG,GAG K228E XP_016879154.1
XM_017023666.1 611 Missense Mutation AAG,GAG K173E XP_016879155.1

View Full Product Details