Product Details

SNP ID
rs190068917
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:110712975 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGAGTGGTGGTCCGGCCGCGGAA[C/T]GGGTAGGTCTCCCGCGCACTCTGCG
Phenotype
MIM: 612800 MIM: 601566
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CARS2 PubMed Links

Gene Details

Gene
CARS2
Gene Name
cysteinyl-tRNA synthetase 2, mitochondrial (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024537.3 353 Intron NP_078813.1
XM_006719953.2 353 Intron XP_006720016.1
XM_011521116.1 353 Intron XP_011519418.1
XM_011521117.2 353 Intron XP_011519419.1
XM_011521118.2 353 Intron XP_011519420.1
XM_017020741.1 353 Intron XP_016876230.1
XM_017020742.1 353 Intron XP_016876231.1
Gene
ING1
Gene Name
inhibitor of growth family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267728.1 353 Intron NP_001254657.1
NM_005537.5 353 Intron NP_005528.4
NM_198217.2 353 Missense Mutation ACG,ATG T1M NP_937860.1
NM_198218.2 353 Intron NP_937861.1
NM_198219.2 353 Intron NP_937862.1
Gene
LOC105376684
Gene Name
translation initiation factor IF-2-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011521137.2 353 UTR 3 XP_011519439.1

View Full Product Details