Product Details

SNP ID
rs190094264
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:93741270 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCAATGTAGGGCCCCGGGAGGGC[A/G]CCTTCGGAGGGGCGTCGCAACCAGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM172A PubMed Links

Gene Details

Gene
FAM172A
Gene Name
family with sequence similarity 172 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163417.1 334 Intron NP_001156889.1
NM_001163418.1 334 Intron NP_001156890.1
NM_032042.5 334 Intron NP_114431.2
XM_005272103.3 334 Intron XP_005272160.1
XM_005272105.3 334 Intron XP_005272162.1
XM_005272106.3 334 Intron XP_005272163.1
XM_005272108.3 334 Intron XP_005272165.1
XM_005272109.4 334 Intron XP_005272166.1
XM_005272110.3 334 Intron XP_005272167.1
XM_005272111.4 334 Intron XP_005272168.1
XM_006714717.3 334 Intron XP_006714780.1
XM_006714718.3 334 Intron XP_006714781.1
XM_006714719.3 334 Intron XP_006714782.1
XM_011543668.2 334 Intron XP_011541970.1
XM_011543670.2 334 Intron XP_011541972.1
XM_011543671.2 334 Intron XP_011541973.1
XM_011543672.2 334 Intron XP_011541974.1
XM_011543673.2 334 Intron XP_011541975.1
XM_011543674.2 334 Intron XP_011541976.1
XM_017009952.1 334 Intron XP_016865441.1
XM_017009953.1 334 Intron XP_016865442.1
XM_017009954.1 334 Intron XP_016865443.1
XM_017009955.1 334 Intron XP_016865444.1
XM_017009956.1 334 Intron XP_016865445.1
XM_017009957.1 334 Intron XP_016865446.1
XM_017009958.1 334 Intron XP_016865447.1
XM_017009959.1 334 Intron XP_016865448.1
XM_017009960.1 334 Intron XP_016865449.1
XM_017009961.1 334 Intron XP_016865450.1
XM_017009962.1 334 Intron XP_016865451.1
Gene
POU5F2
Gene Name
POU domain class 5, transcription factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153216.1 334 Silent Mutation GGC,GGT G98G NP_694948.1

View Full Product Details