Product Details

SNP ID
rs190478032
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:51789588 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCACATGAACGCTGAGCATTTTAC[A/T]TCCTGGCCCTCTATGGGCCTTGAAC
Phenotype
MIM: 602651 MIM: 606737
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
NRDC PubMed Links

Gene Details

Gene
NRDC
Gene Name
nardilysin convertase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101662.1 3560 Missense Mutation AGT,TGT S1080C NP_001095132.1
NM_001242361.1 3560 Missense Mutation AGT,TGT S1016C NP_001229290.1
NM_002525.2 3560 Missense Mutation AGT,TGT S1148C NP_002516.2
XM_005270903.1 3560 Missense Mutation AGT,TGT S843C XP_005270960.1
XM_011541522.1 3560 Missense Mutation AGT,TGT S1038C XP_011539824.1
XM_011541523.1 3560 Missense Mutation AGT,TGT S948C XP_011539825.1
XM_011541525.2 3560 Missense Mutation AGT,TGT S843C XP_011539827.1
XM_017001375.1 3560 Missense Mutation AGT,TGT S948C XP_016856864.1
Gene
OSBPL9
Gene Name
oxysterol binding protein like 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024586.5 3560 Intron NP_078862.4
NM_148904.3 3560 Intron NP_683702.1
NM_148905.3 3560 Intron NP_683703.1
NM_148906.2 3560 Intron NP_683704.2
NM_148907.2 3560 Intron NP_683705.1
NM_148908.3 3560 Intron NP_683706.3
NM_148909.3 3560 Intron NP_683707.3
XM_006710318.2 3560 Intron XP_006710381.1
XM_006710320.3 3560 Intron XP_006710383.1
XM_006710321.2 3560 Intron XP_006710384.1
XM_006710322.3 3560 Intron XP_006710385.1
XM_006710323.2 3560 Intron XP_006710386.1
XM_006710324.3 3560 Intron XP_006710387.1
XM_006710325.3 3560 Intron XP_006710388.1
XM_006710326.2 3560 Intron XP_006710389.1
XM_011540599.2 3560 Intron XP_011538901.1
XM_011540600.2 3560 Intron XP_011538902.1
XM_011540601.2 3560 Intron XP_011538903.1
XM_011540602.2 3560 Intron XP_011538904.1
XM_011540603.2 3560 Intron XP_011538905.1
XM_011540604.2 3560 Intron XP_011538906.1
XM_017000218.1 3560 Intron XP_016855707.1
XM_017000219.1 3560 Intron XP_016855708.1
XM_017000220.1 3560 Intron XP_016855709.1
XM_017000221.1 3560 Intron XP_016855710.1
XM_017000222.1 3560 Intron XP_016855711.1
XM_017000223.1 3560 Intron XP_016855712.1
XM_017000224.1 3560 Intron XP_016855713.1
XM_017000225.1 3560 Intron XP_016855714.1

View Full Product Details