Product Details

SNP ID
rs190306578
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:36169900 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTAAGTCCATCTGCCGTGCCAACT[G/T]CTTGCGCTACCTCTTCTTGGCTGAG
Phenotype
MIM: 603960 MIM: 607141
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCIN PubMed Links

Gene Details

Gene
CCIN
Gene Name
calicin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005893.2 509 Missense Mutation TGC,TTC C133F NP_005884.2
Gene
GLIPR2
Gene Name
GLI pathogenesis related 2
There are no transcripts associated with this gene.

View Full Product Details