Product Details

SNP ID
rs190610162
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:21407525 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGATGGTCTCTATCTCCTGACCTC[A/G]TGATCCGCCCGCCTCAGTCTCCCAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF493 PubMed Links

Gene Details

Gene
ZNF493
Gene Name
zinc finger protein 493
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001076678.2 428 Intron NP_001070146.1
NM_145326.2 428 Silent Mutation TCA,TCG S103S NP_663299.2
NM_175910.6 428 Intron NP_787106.4

View Full Product Details