Product Details

SNP ID
rs1042579279
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:103086613 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCTATACATTCTTCAAGGTCAAG[C/T]TTAAATGATCCTTCTTCCATAAGGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GIN1 PubMed Links

Gene Details

Gene
GIN1
Gene Name
gypsy retrotransposon integrase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317954.1 2742 UTR 3 NP_001304883.1
NM_017676.2 2742 UTR 3 NP_060146.2

View Full Product Details