Product Details
- SNP ID
-
rs190715442
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:47166513 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCCCACCAGGGCCCTACCTGCTCC[A/G]TGAGCTCCATCCATGTCCCCAAAGA
- Phenotype
-
MIM: 606908
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ARFGAP2
PubMed Links
Gene Details
- Gene
- ARFGAP2
- Gene Name
- ADP ribosylation factor GTPase activating protein 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001242832.1 |
2144 |
Silent Mutation |
CAC,CAT |
H445H |
NP_001229761.1 |
NM_032389.4 |
2144 |
Silent Mutation |
CAC,CAT |
H473H |
NP_115765.2 |
XM_005253166.1 |
2144 |
Silent Mutation |
CAC,CAT |
H512H |
XP_005253223.1 |
XM_005253167.1 |
2144 |
Silent Mutation |
CAC,CAT |
H501H |
XP_005253224.1 |
XM_005253168.2 |
2144 |
Silent Mutation |
CAC,CAT |
H487H |
XP_005253225.1 |
XM_006718346.1 |
2144 |
Silent Mutation |
CAC,CAT |
H498H |
XP_006718409.1 |
XM_011520405.1 |
2144 |
Silent Mutation |
CAC,CAT |
H459H |
XP_011518707.1 |
XM_017018413.1 |
2144 |
Silent Mutation |
CAC,CAT |
H487H |
XP_016873902.1 |
XM_017018414.1 |
2144 |
Silent Mutation |
CAC,CAT |
H350H |
XP_016873903.1 |
XM_017018415.1 |
2144 |
Silent Mutation |
CAC,CAT |
H336H |
XP_016873904.1 |
- Gene
- C11orf49
- Gene Name
- chromosome 11 open reading frame 49
There are no transcripts associated with this gene.
View Full Product Details