Product Details

SNP ID
rs190796582
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:1232416 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCCAGGGCGATGTCGGGCCGCAG[C/T]CCGCCTCCCCCCGCGCCCGCGCGCG
Phenotype
MIM: 615291 MIM: 614282
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
B3GALT6 PubMed Links

Gene Details

Gene
B3GALT6
Gene Name
beta-1,3-galactosyltransferase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080605.3 168 Silent Mutation AGC,AGT S46S NP_542172.2
Gene
SDF4
Gene Name
stromal cell derived factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016176.3 168 Intron NP_057260.2
NM_016547.2 168 Intron NP_057631.1
XM_011541556.1 168 Intron XP_011539858.1

View Full Product Details