Product Details

SNP ID
rs190803760
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:11994096 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCCATTTTTATAGCATTTCTTGT[G/T]CATTTCTGATGATAATGAATTTATA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DEFB134 PubMed Links

Gene Details

Gene
DEFB134
Gene Name
defensin beta 134
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302695.1 177 Missense Mutation AAC,CAC N29H NP_001289624.1
XM_017013724.1 177 Missense Mutation AAC,CAC N29H XP_016869213.1
Gene
DEFB135
Gene Name
defensin beta 135
There are no transcripts associated with this gene.

View Full Product Details