Product Details

SNP ID
rs191174298
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49118734 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTACCCTTAAGAGCGTCTGAGGGC[A/G]AAGGCCAAGTCCCTTCCTCCAGAGT
Phenotype
MIM: 612331 MIM: 603144
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
C19orf73 PubMed Links
Additional Information
For this assay, SNP(s) [rs2232003] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
C19orf73
Gene Name
chromosome 19 open reading frame 73
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018111.2 407 Missense Mutation CGC,TGC R97C NP_060581.2
Gene
LIN7B
Gene Name
lin-7 homolog B, crumbs cell polarity complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308419.1 407 Intron NP_001295348.1
NM_022165.2 407 Intron NP_071448.1
XM_006723323.3 407 Intron XP_006723386.1
XM_017027131.1 407 Intron XP_016882620.1
Gene
PPFIA3
Gene Name
PTPRF interacting protein alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003660.3 407 Intron NP_003651.1
XM_017027407.1 407 Intron XP_016882896.1

View Full Product Details