Product Details

SNP ID
rs191203230
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:79363028 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAATGATTGCAATGTTGTCATCCT[C/T]ACGAATGTCAGCCTCCTCAGTCACA
Phenotype
MIM: 300222
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ITM2A PubMed Links

Gene Details

Gene
ITM2A
Gene Name
integral membrane protein 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171581.1 692 Missense Mutation AAG,GAG K75E NP_001165052.1
NM_004867.4 692 Missense Mutation AAG,GAG K119E NP_004858.1

View Full Product Details