Product Details

SNP ID
rs191612367
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:33379270 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGATATCGCAGCAAAACAGCACT[C/G]CAGGGGTGAACGGAATTAGTGTTAT
Phenotype
MIM: 138972 MIM: 613230
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CEBPG PubMed Links

Gene Details

Gene
CEBPG
Gene Name
CCAAT/enhancer binding protein gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252296.1 217 Missense Mutation CCA,GCA P11A NP_001239225.1
NM_001806.3 217 Missense Mutation CCA,GCA P11A NP_001797.1
Gene
PEPD
Gene Name
peptidase D
There are no transcripts associated with this gene.

View Full Product Details