Product Details

SNP ID
rs191255872
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:122208560 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGAATGTGATTCCTGGCGGTTA[G/T]AGAGGCCTGATCTGCGCCTGCCAAA
Phenotype
MIM: 605864 MIM: 605219
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
B3GNT4 PubMed Links

Gene Details

Gene
B3GNT4
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011538732.1 934 Intron XP_011537034.1
Gene
DIABLO
Gene Name
diablo IAP-binding mitochondrial protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278302.1 934 Missense Mutation ATA,CTA I84L NP_001265231.1
NM_001278303.1 934 Missense Mutation ATA,CTA I108L NP_001265232.1
NM_001278304.1 934 Missense Mutation ATA,CTA I128L NP_001265233.1
NM_001278342.1 934 Missense Mutation ATA,CTA I137L NP_001265271.1
NM_019887.5 934 Missense Mutation ATA,CTA I181L NP_063940.1
NM_138930.3 934 Missense Mutation ATA,CTA I128L NP_620308.1
Gene
LRRC43
Gene Name
leucine rich repeat containing 43
There are no transcripts associated with this gene.

View Full Product Details