Product Details
- SNP ID
-
rs191277814
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:20574386 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACTGCCAGTTTTTCCTAGGTACAA[A/G]TGATGCAAAATGTGCACCTGGCTCC
- Phenotype
-
MIM: 600595
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IFT88
PubMed Links
Gene Details
- Gene
- IFT88
- Gene Name
- intraflagellar transport 88
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001318491.1 |
1156 |
Missense Mutation |
ATG,GTG |
M1V |
NP_001305420.1 |
NM_001318493.1 |
1156 |
Missense Mutation |
ATG,GTG |
M10V |
NP_001305422.1 |
NM_006531.4 |
1156 |
Missense Mutation |
ATG,GTG |
M1V |
NP_006522.2 |
NM_175605.4 |
1156 |
Missense Mutation |
ATG,GTG |
M10V |
NP_783195.2 |
XM_005266553.2 |
1156 |
Missense Mutation |
ATG,GTG |
M10V |
XP_005266610.1 |
XM_006719870.3 |
1156 |
Missense Mutation |
ATG,GTG |
M1V |
XP_006719933.1 |
XM_011535241.2 |
1156 |
Missense Mutation |
ATG,GTG |
M10V |
XP_011533543.1 |
XM_011535242.1 |
1156 |
Missense Mutation |
ATG,GTG |
M1V |
XP_011533544.1 |
XM_011535243.1 |
1156 |
Missense Mutation |
ATG,GTG |
M1V |
XP_011533545.1 |
XM_017020757.1 |
1156 |
Missense Mutation |
ATG,GTG |
M1V |
XP_016876246.1 |
XM_017020758.1 |
1156 |
Missense Mutation |
ATG,GTG |
M10V |
XP_016876247.1 |
XM_017020759.1 |
1156 |
Missense Mutation |
ATG,GTG |
M10V |
XP_016876248.1 |
XM_017020760.1 |
1156 |
Missense Mutation |
ATG,GTG |
M10V |
XP_016876249.1 |
XM_017020761.1 |
1156 |
Missense Mutation |
ATG,GTG |
M10V |
XP_016876250.1 |
XM_017020762.1 |
1156 |
Missense Mutation |
ATG,GTG |
M10V |
XP_016876251.1 |
XM_017020763.1 |
1156 |
Missense Mutation |
ATG,GTG |
M10V |
XP_016876252.1 |
XM_017020764.1 |
1156 |
Missense Mutation |
ATG,GTG |
M1V |
XP_016876253.1 |
XM_017020765.1 |
1156 |
Missense Mutation |
ATG,GTG |
M1V |
XP_016876254.1 |
XM_017020766.1 |
1156 |
Missense Mutation |
ATG,GTG |
M1V |
XP_016876255.1 |
XM_017020767.1 |
1156 |
Missense Mutation |
ATG,GTG |
M1V |
XP_016876256.1 |
XM_017020768.1 |
1156 |
Missense Mutation |
ATG,GTG |
M10V |
XP_016876257.1 |
XM_017020769.1 |
1156 |
Missense Mutation |
ATG,GTG |
M1V |
XP_016876258.1 |
XM_017020770.1 |
1156 |
Missense Mutation |
ATG,GTG |
M10V |
XP_016876259.1 |
XM_017020771.1 |
1156 |
Missense Mutation |
ATG,GTG |
M10V |
XP_016876260.1 |
XM_017020772.1 |
1156 |
Missense Mutation |
ATG,GTG |
M1V |
XP_016876261.1 |
XM_017020773.1 |
1156 |
Missense Mutation |
ATG,GTG |
M1V |
XP_016876262.1 |
XM_017020774.1 |
1156 |
UTR 5 |
|
|
XP_016876263.1 |
XM_017020775.1 |
1156 |
Missense Mutation |
ATG,GTG |
M10V |
XP_016876264.1 |
XM_017020776.1 |
1156 |
Missense Mutation |
ATG,GTG |
M10V |
XP_016876265.1 |
View Full Product Details