Product Details

SNP ID
rs191285090
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:7651112 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGCTTCATCCCCAGGTGGGTAGT[C/T]GACAAAATTCCTCCAGCAGTGATAA
Phenotype
MIM: 600130
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APOBEC1 PubMed Links

Gene Details

Gene
APOBEC1
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304566.1 496 Missense Mutation AAC,GAC N158D NP_001291495.1
NM_001644.4 496 Missense Mutation AAC,GAC N158D NP_001635.2
NM_005889.3 496 Missense Mutation AAC,GAC N113D NP_005880.2

View Full Product Details