Product Details

SNP ID
rs191391350
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42023824 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTCTACTTGGCCAGCAAGATGAC[A/G]CAACCCCAGAGCAAGTCTGCCTTCC
Phenotype
MIM: 601964 MIM: 604497
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJC7 PubMed Links

Gene Details

Gene
DNAJC7
Gene Name
DnaJ heat shock protein family (Hsp40) member C7
There are no transcripts associated with this gene.

Gene
NKIRAS2
Gene Name
NFKB inhibitor interacting Ras like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001349.2 572 Silent Mutation ACA,ACG T169T NP_001001349.1
NM_001144927.1 572 Silent Mutation ACA,ACG T169T NP_001138399.1
NM_001144928.1 572 Silent Mutation ACA,ACG T113T NP_001138400.1
NM_001144929.1 572 UTR 3 NP_001138401.1
NM_017595.5 572 Silent Mutation ACA,ACG T169T NP_060065.2
XM_005257251.1 572 Silent Mutation ACA,ACG T169T XP_005257308.1
Gene
ZNF385C
Gene Name
zinc finger protein 385C
There are no transcripts associated with this gene.

View Full Product Details