Product Details
- SNP ID
-
rs191110743
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:76429160 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTCCGTGCTGCCTAACCCTGGTGA[A/G]GGCCCGAGCACATTGTTTAGCTCTA
- Phenotype
-
MIM: 300759
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MAGEE1
PubMed Links
Gene Details
- Gene
- MAGEE1
- Gene Name
- MAGE family member E1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020932.2 |
1508 |
Silent Mutation |
GAA,GAG |
E410E |
NP_065983.1 |
View Full Product Details