Product Details

SNP ID
rs191143712
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.21:44637739 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGTGCCCGTCTGCTGCGTGCCC[A/G]TCTGTAACAAGCCTGTGTGCTTCGT
Phenotype
MIM: 612920
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
KRTAP10-10 PubMed Links
Additional Information
For this assay, SNP(s) [rs4818947] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
KRTAP10-10
Gene Name
keratin associated protein 10-10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181688.2 Intron NP_859016.1
Gene
KRTAP10-11
Gene Name
keratin associated protein 10-11
There are no transcripts associated with this gene.

Gene
KRTAP10-9
Gene Name
keratin associated protein 10-9
There are no transcripts associated with this gene.

Gene
TSPEAR
Gene Name
thrombospondin type laminin G domain and EAR repeats
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272037.1 Intron NP_001258966.1
NM_144991.2 Intron NP_659428.2

View Full Product Details