Product Details
- SNP ID
-
rs191729623
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:27435884 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCATAGTCTGGAGTATTTGATGAA[A/C]TATCTCCTCCTAAAAAAGGGAAACA
- Phenotype
-
MIM: 600860
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
GTF3A
PubMed Links
Gene Details
- Gene
- GTF3A
- Gene Name
- general transcription factor IIIA
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002097.2 |
1516 |
Intron |
|
|
NP_002088.2 |
- Gene
- MTIF3
- Gene Name
- mitochondrial translational initiation factor 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001166261.1 |
1516 |
Intron |
|
|
NP_001159733.1 |
NM_001166262.1 |
1516 |
Intron |
|
|
NP_001159734.1 |
NM_001166263.1 |
1516 |
Missense Mutation |
GTT,TTT |
V210F |
NP_001159735.1 |
NM_152912.4 |
1516 |
Intron |
|
|
NP_690876.3 |
XM_006719771.3 |
1516 |
Missense Mutation |
GTT,TTT |
V210F |
XP_006719834.1 |
XM_006719772.3 |
1516 |
Missense Mutation |
GTT,TTT |
V210F |
XP_006719835.1 |
XM_011534957.2 |
1516 |
Missense Mutation |
GTT,TTT |
V210F |
XP_011533259.1 |
XM_011534958.2 |
1516 |
Missense Mutation |
GTT,TTT |
V210F |
XP_011533260.1 |
XM_011534959.2 |
1516 |
Missense Mutation |
GTT,TTT |
V210F |
XP_011533261.1 |
XM_011534960.2 |
1516 |
Missense Mutation |
GTT,TTT |
V210F |
XP_011533262.1 |
XM_011534961.2 |
1516 |
Missense Mutation |
GTT,TTT |
V210F |
XP_011533263.1 |
XM_011534962.2 |
1516 |
Missense Mutation |
GTT,TTT |
V210F |
XP_011533264.1 |
XM_011534963.2 |
1516 |
Missense Mutation |
GTT,TTT |
V210F |
XP_011533265.1 |
XM_017020417.1 |
1516 |
Intron |
|
|
XP_016875906.1 |
View Full Product Details