Product Details

SNP ID
rs192010535
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:91908672 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCGCGCGGCGAGCGGCGTAGGCTG[A/C]GCAGCGCTCCGGATGCCCGCGGTAG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FGFBP3 PubMed Links

Gene Details

Gene
FGFBP3
Gene Name
fibroblast growth factor binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152429.4 480 Missense Mutation GCA,TCA A100S NP_689642.3

View Full Product Details