Product Details

SNP ID
rs192169308
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:39966329 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCGAGTGGAGGGAGATGTGGATG[C/T]ACCAAAGACGGTGAAGGTGAAGGGA
Phenotype
MIM: 611218
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
GSDMA PubMed Links

Gene Details

Gene
GSDMA
Gene Name
gasdermin A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178171.4 402 Missense Mutation GCA,GTA A95V NP_835465.2
XM_006721832.2 402 Missense Mutation GCA,GTA A95V XP_006721895.1
XM_011524651.2 402 Intron XP_011522953.1
XM_017024502.1 402 Missense Mutation GCA,GTA A95V XP_016879991.1
XM_017024503.1 402 Missense Mutation GCA,GTA A95V XP_016879992.1
XM_017024504.1 402 Missense Mutation GCA,GTA A95V XP_016879993.1

View Full Product Details