Product Details

SNP ID
rs191858058
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:59269020 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGTGCCCCTAGAGGGCCTGCCAC[G/T]GCTTCCCGATGGCCTGGATGTGCTC
Phenotype
MIM: 601881
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RAX PubMed Links

Gene Details

Gene
RAX
Gene Name
retina and anterior neural fold homeobox
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013435.2 1212 Missense Mutation CAG,CCG Q342P NP_038463.2

View Full Product Details