Product Details

SNP ID
rs191672938
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30590319 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCATATCCAGAGTTACCATATTCA[G/T]AGCCATAGCTGCAACCATAGTCCAG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KRTAP22-2 PubMed Links

Gene Details

Gene
KRTAP22-2
Gene Name
keratin associated protein 22-2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164434.1 79 Missense Mutation TAT,TCT Y19S NP_001157906.1
Gene
KRTAP6-2
Gene Name
keratin associated protein 6-2
There are no transcripts associated with this gene.

Gene
KRTAP6-3
Gene Name
keratin associated protein 6-3
There are no transcripts associated with this gene.

View Full Product Details