Product Details

SNP ID
rs191662436
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44360051 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGGTGCCAGCTGCCAGCAGAGTC[C/T]GGTTCCCAAACACATCCACTGAGGC
Phenotype
MIM: 138945
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM171A2 PubMed Links

Gene Details

Gene
FAM171A2
Gene Name
family with sequence similarity 171 member A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198475.2 353 Missense Mutation CAG,CGG Q67R NP_940877.2
XM_017024490.1 353 Intron XP_016879979.1
Gene
GRN
Gene Name
granulin
There are no transcripts associated with this gene.

View Full Product Details