Product Details

SNP ID
rs192595388
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:76623319 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGACCTCTCTCATAGCTTTTCTTT[C/T]GCAGGTATTCCTAGCTCTTCTTACA
Phenotype
MIM: 610138
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ST6GALNAC1 PubMed Links

Gene Details

Gene
ST6GALNAC1
Gene Name
ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289107.1 Intron NP_001276036.1
NM_018414.4 Intron NP_060884.1
XM_011524995.2 Intron XP_011523297.1
XM_011524996.2 Intron XP_011523298.1
XM_017024842.1 Intron XP_016880331.1
XM_017024843.1 Intron XP_016880332.1
XM_017024844.1 Intron XP_016880333.1

View Full Product Details