Product Details

SNP ID
rs193079087
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:8823207 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACTACCTGGTGACATTACCAGCC[C/T]GGCTAAATTTCCCCTCCGTTCAGAA
Phenotype
MIM: 610627
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
A2ML1 PubMed Links

Gene Details

Gene
A2ML1
Gene Name
alpha-2-macroglobulin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282424.2 186 Intron NP_001269353.1
NM_144670.5 186 Missense Mutation CGG,TGG R30W NP_653271.2
XM_011520566.2 186 Missense Mutation CGG,TGG R30W XP_011518868.1
XM_011520567.2 186 Missense Mutation CGG,TGG R30W XP_011518869.1
XM_017018868.1 186 Missense Mutation CGG,TGG R30W XP_016874357.1
XM_017018869.1 186 Missense Mutation CGG,TGG R30W XP_016874358.1
XM_017018870.1 186 Missense Mutation CGG,TGG R30W XP_016874359.1

View Full Product Details