Product Details

SNP ID
rs192855237
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:5302484 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGGCATAGCACAGGGTCATGGT[A/G]ACCACAGGGCCCACGAACTCGACAC
Phenotype
MIM: 607123
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PROKR2 PubMed Links

Gene Details

Gene
PROKR2
Gene Name
prokineticin receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144773.3 958 Silent Mutation GTC,GTT V237V NP_658986.1
XM_011529159.2 958 Silent Mutation GTC,GTT V201V XP_011527461.1
XM_017027646.1 958 Silent Mutation GTC,GTT V237V XP_016883135.1

View Full Product Details