Product Details

SNP ID
rs192871611
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:99648197 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACATGGAGGACTGCTGTATGCTAC[C/T]GTATTACACGGCCCAAAGCAGCCCC
Phenotype
MIM: 607815
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKS1B PubMed Links

Gene Details

Gene
ANKS1B
Gene Name
ankyrin repeat and sterile alpha motif domain containing 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204065.1 448 Intron NP_001190994.1
NM_001204066.1 448 Intron NP_001190995.1
NM_001204067.1 448 Intron NP_001190996.1
NM_001204068.1 448 Intron NP_001190997.1
NM_001204069.1 448 Intron NP_001190998.1
NM_001204070.1 448 Intron NP_001190999.1
NM_001204079.1 448 Intron NP_001191008.1
NM_001204080.1 448 Intron NP_001191009.1
NM_001204081.1 448 Intron NP_001191010.1
NM_020140.3 448 Intron NP_064525.1
NM_152788.4 448 Intron NP_690001.3
NM_181670.3 448 Intron NP_858056.2
XM_005269028.4 448 Intron XP_005269085.1
XM_005269029.4 448 Intron XP_005269086.1
XM_005269032.3 448 Intron XP_005269089.1
XM_006719504.3 448 Intron XP_006719567.1
XM_006719505.3 448 Intron XP_006719568.1
XM_006719506.3 448 Intron XP_006719569.1
XM_006719507.3 448 Intron XP_006719570.1
XM_006719508.3 448 Intron XP_006719571.1
XM_006719509.3 448 Intron XP_006719572.1
XM_006719510.3 448 Intron XP_006719573.1
XM_006719512.3 448 Intron XP_006719575.1
XM_006719513.3 448 Intron XP_006719576.1
XM_006719514.3 448 Intron XP_006719577.1
XM_011538571.2 448 Intron XP_011536873.1
XM_017019651.1 448 Intron XP_016875140.1
XM_017019652.1 448 Intron XP_016875141.1
XM_017019653.1 448 Intron XP_016875142.1
XM_017019654.1 448 Intron XP_016875143.1
XM_017019655.1 448 Intron XP_016875144.1
XM_017019656.1 448 Intron XP_016875145.1
XM_017019657.1 448 Intron XP_016875146.1
XM_017019658.1 448 Intron XP_016875147.1
XM_017019659.1 448 Intron XP_016875148.1
XM_017019660.1 448 Intron XP_016875149.1
XM_017019661.1 448 Intron XP_016875150.1
XM_017019662.1 448 Intron XP_016875151.1
XM_017019663.1 448 Intron XP_016875152.1
XM_017019664.1 448 Intron XP_016875153.1
XM_017019665.1 448 Intron XP_016875154.1
Gene
FAM71C
Gene Name
family with sequence similarity 71 member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153364.3 448 Missense Mutation CCG,CTG P8L NP_699195.1

View Full Product Details