Product Details

SNP ID
rs192854934
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56743140 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAAATCAGGATAGCAAGAATGCT[A/G]GAGAGTACTGTGAAGCCCACCACAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR6128 PubMed Links

Gene Details

Gene
MIR6128
Gene Name
microRNA 6128
There are no transcripts associated with this gene.

Gene
OR9G4
Gene Name
olfactory receptor family 9 subfamily G member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005284.1 672 Silent Mutation TCC,TCT S224S NP_001005284.1

View Full Product Details