Product Details

SNP ID
rs192883326
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:150491485 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCTTCCGGAGGCTAGAGGCTTCA[C/T]GGGATCAGCTTCGCCAGTTGTTCAA
Phenotype
MIM: 612805
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR6878 PubMed Links

Gene Details

Gene
MIR6878
Gene Name
microRNA 6878
There are no transcripts associated with this gene.

Gene
TARS2
Gene Name
threonyl-tRNA synthetase 2, mitochondrial (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271895.1 654 Missense Mutation CGG,TGG R202W NP_001258824.1
NM_001271896.1 654 Missense Mutation CGG,TGG R202W NP_001258825.1
NM_025150.4 654 Missense Mutation CGG,TGG R202W NP_079426.2
XM_006711555.1 654 Missense Mutation CGG,TGG R202W XP_006711618.1
XM_006711556.1 654 Missense Mutation CGG,TGG R202W XP_006711619.1
XM_017002394.1 654 Missense Mutation CGG,TGG R202W XP_016857883.1
XM_017002395.1 654 Missense Mutation CGG,TGG R202W XP_016857884.1

View Full Product Details