Product Details

SNP ID
rs192981266
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:47802776 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTTCTGGGCTCTGCACCCAGGGC[C/T]ACCCCAGGCCTGGTGGAGGCTGCAG
Phenotype
MIM: 611166
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TPRX1 PubMed Links

Gene Details

Gene
TPRX1
Gene Name
tetrapeptide repeat homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198479.2 306 Missense Mutation AGC,GGC S79G NP_940881.2

View Full Product Details