Product Details

SNP ID
rs192725460
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:69957619 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGGACAGGCATCACTCTGCCCTT[A/T]GTGTAGACAGTGTCTTTAGCTTTAG
Phenotype
MIM: 115460
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CSN2 PubMed Links

Gene Details

Gene
CSN2
Gene Name
casein beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302770.1 346 Silent Mutation ACA,ACT T109T NP_001289699.1
NM_001891.3 346 Silent Mutation ACA,ACT T110T NP_001882.1
XM_017007760.1 346 Silent Mutation ACA,ACT T110T XP_016863249.1

View Full Product Details