Product Details

SNP ID
rs193279617
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:59210164 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGGAGGGAGCGCGGCTGGCGGAC[A/C]GGAGCCTCCATGGCGGGAGGATGAG
Phenotype
MIM: 615920 MIM: 613175
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PRR11 PubMed Links

Gene Details

Gene
PRR11
Gene Name
proline rich 11
There are no transcripts associated with this gene.

Gene
SMG8
Gene Name
SMG8, nonsense mediated mRNA decay factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018149.6 155 Missense Mutation CAG,CCG Q38P NP_060619.4

View Full Product Details