Product Details

SNP ID
rs340656
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:6866361 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGCTTTTTTTTTTTTCTTTAAAA[A/G]TGTAGTTTCTAAAAGTCTGTAATCT
Phenotype
MIM: 604101
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRM7 PubMed Links

Gene Details

Gene
GRM7
Gene Name
glutamate metabotropic receptor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000844.3 Intron NP_000835.1
NM_181874.2 Intron NP_870989.1
XM_017006272.1 Intron XP_016861761.1
XM_017006273.1 Intron XP_016861762.1

View Full Product Details