Product Details

SNP ID
rs5992970
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:18031539 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATAACCAACATTAGTATTTTACT[C/T]AGAACATTAAGCACTGTTCTAGGTG
Phenotype
MIM: 608882
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FLJ41941 PubMed Links

Gene Details

Gene
FLJ41941
Gene Name
uncharacterized LOC100192420
There are no transcripts associated with this gene.

Gene
MICAL3
Gene Name
microtubule associated monooxygenase, calponin and LIM domain containing 3
There are no transcripts associated with this gene.

View Full Product Details