Product Details

SNP ID
rs6124568
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:43508044 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGGAGGCTGCGTGTTTGGAGATA[A/C]GCAATCAATTAAGAGTTTTTTTATT
Phenotype
MIM: 608802
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
L3MBTL1 PubMed Links

Gene Details

Gene
L3MBTL1
Gene Name
l(3)mbt-like 1 (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015478.6 Intron NP_056293.4
NM_032107.4 Intron NP_115479.4

View Full Product Details