Product Details

SNP ID
rs6669931
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:212437015 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGATGGTGGTGTGCACAGGCTTAGC[A/G]TAGGCCTGGAATAGGGCTTAGCACC
Phenotype
MIM: 611874
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NENF PubMed Links

Gene Details

Gene
NENF
Gene Name
neudesin neurotrophic factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013349.4 Intron NP_037481.1

View Full Product Details