Product Details
- SNP ID
-
rs718721
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:32512134 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGAAACCTTTGAACATATTCTATGA[C/T]TCTATGAAAAAGAGGATCCAGATAT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
WBP11P1
PubMed Links
Gene Details
- Gene
- WBP11P1
- Gene Name
- WW domain binding protein 11 pseudogene 1
There are no transcripts associated with this gene.
View Full Product Details