Product Details

SNP ID
rs7160061
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:49601205 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTAACCTAAATATTTCTATATTAA[A/G]GCTTAATGTGCTTTCTTAAAAAAAA
Phenotype
MIM: 609193
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100288910 PubMed Links

Gene Details

Gene
LOC100288910
Gene Name
uncharacterized LOC100288910
There are no transcripts associated with this gene.

Gene
LRR1
Gene Name
leucine rich repeat protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152329.3 Intron NP_689542.2
NM_203467.1 Intron NP_982292.1

View Full Product Details