Product Details
- SNP ID
-
hCV189280487
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:19345382 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTATTGATATTTCCTCACCCTTCGG[C/T]TCTCAGTAAAGGATGGTTTTTCCAC
- Phenotype
-
MIM: 600237
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HIRA
PubMed Links
Gene Details
- Gene
- HIRA
- Gene Name
- histone cell cycle regulator
There are no transcripts associated with this gene.
View Full Product Details