Product Details

SNP ID
rs74234036
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:33077935 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATGGATTCTCAGTCCTGGTGTTT[A/T]TAAAAATCTCATTCCAGCCGGGCGC
Phenotype
MIM: 600367 MIM: 612296
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CSTF3 PubMed Links

Gene Details

Gene
CSTF3
Gene Name
cleavage stimulation factor subunit 3
There are no transcripts associated with this gene.

Gene
LINC00294
Gene Name
long intergenic non-protein coding RNA 294
There are no transcripts associated with this gene.

Gene
TCP11L1
Gene Name
t-complex 11 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145541.1 7203 Intron NP_001139013.1
NM_018393.3 7203 Intron NP_060863.3
XM_011520204.2 7203 UTR 3 XP_011518506.1
XM_011520205.2 7203 UTR 3 XP_011518507.1
XM_011520206.2 7203 UTR 3 XP_011518508.1
XM_011520207.2 7203 UTR 3 XP_011518509.1
XM_017017989.1 7203 UTR 3 XP_016873478.1
XM_017017990.1 7203 UTR 3 XP_016873479.1
XM_017017991.1 7203 UTR 3 XP_016873480.1
XM_017017992.1 7203 UTR 3 XP_016873481.1

View Full Product Details